Methyl 6-chloro-2-methylnicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 6-chloro-2-methylpyridine-3-carboxylate

28181-1G 28181-25G 28181-500MG 28181-5G
76939-312EA 173.99 USD
76939-312 76939-314 76939-316 76939-318
Methyl 6-chloro-2-methylnicotinate ≥97%
Methyl 6-chloro-2-methylnicotinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD14155903
CAS Number: 851759-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR