7-Iodo-1,2,3,4-tetrahydroquinoline ≥97%

Supplier: AOBChem USA

Warning

28086-1G 28086-25G 28086-500MG 28086-5G
76939-284EA 379.81 USD
76939-284 76939-286 76939-288 76939-290
7-Iodo-1,2,3,4-tetrahydroquinoline ≥97%
7-Iodo-1,2,3,4-tetrahydroquinoline

Precaución: for Research Use Only

Formula: C₉H₁₀IN
Storage Temperature: Ambient
MDL Number: MFCD08544274
CAS Number: 939758-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR