Ethyl-5-bromopicolinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 5-bromopicolinate, Ethyl 5-bromopyridine-2-carboxylate

28084-25G 28084-5G
76939-272EA 381.93 USD
76939-272 76939-274
Ethyl-5-bromopicolinate ≥97%
Ethyl 5-bromopicolinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD00234415
CAS Number: 77199-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR