Methyl-2,4-dichloronicotinate ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: Methyl 2,4-dichloronicotinate

27897-25G 27897-5G
76939-248EA 396.78 USD
76939-248 76939-250
Methyl-2,4-dichloronicotinate ≥97% research grade
Methyl-2,4-dichloronicotinate
Formula: C₇H₅Cl₂NO₂
MW: 206.022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100222
CAS Number: 442903-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR