6-Bromo-5-methylimidazo[1,2-a]pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
27823-1G
27823-25G
27823-500MG
27823-5G
76939-226EA
147.47
USD
76939-226
76939-228
76939-230
76939-232
6-Bromo-5-methylimidazo[1,2-a]pyridine ≥97%
6-Bromo-5-methylimidazo[1,2-a]pyridine
Formula:
C₈H₇BrN₂ MW: 211.05 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD20489149 CAS Number: 116355-19-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...