N-(4-Bromo-3-(trifluoromethoxy)phenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

27706-1G 27706-25G 27706-500MG 27706-5G
76939-196EA 183.54 USD
76939-196 76939-198 76939-200 76939-202
N-(4-Bromo-3-(trifluoromethoxy)phenyl)acetamide ≥97%
N-(4-Bromo-3-(trifluoromethoxy)phenyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₇BrF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD28681176
CAS Number: 1426805-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR