2-(9-Anthryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97

Supplier: AOBChem USA

Warning

Synonyms: 9-Anthraceneboronic acid pinacol ester, 2-(Anthracen-9-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

27622-25G 27622-5G
76939-178EA 630.18 USD
76939-178 76939-180
2-(9-Anthryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97
2-(9-Anthryl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₂₀H₂₁BO₂
MW: 304.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24039741
CAS Number: 709022-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR