Methyl-2-bromo-6-methoxybenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
Methyl 2-bromo-6-methoxybenzoate, 2-Bromo-6-methoxybenzoic acid methyl ester
27536-1G
27536-25G
27536-500MG
27536-5G
76939-164EA
201.58
USD
76939-164
76939-166
76939-168
76939-170
Methyl-2-bromo-6-methoxybenzoate ≥97%
Methyl-2-bromo-6-methoxybenzoate
Precaución: for Research Use Only
Formula:
C₉H₉BrO₃ MW: 245.07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14697992 CAS Number: 31786-46-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...