Methyl 4-bromo-3-chloro-2-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

27508-1G 27508-25G 27508-500MG 27508-5G
76939-152EA 350.1 USD
76939-152 76939-154 76939-156 76939-158
Methyl 4-bromo-3-chloro-2-fluorobenzoate ≥97%
Methyl 4-bromo-3-chloro-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrClFO₂
MW: 267.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23707534
CAS Number: 1427440-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR