Methyl 3-bromo-2-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

27487-1G 27487-25G 27487-500MG 27487-5G
76939-142EA 162.32 USD
76939-142 76939-144 76939-146 76939-148
Methyl 3-bromo-2-methoxybenzoate ≥97%
Methyl 3-bromo-2-methoxybenzoate
Formula: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD11042778
CAS Number: 260806-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR