6-Bromo-2-chloronicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

27485-1G 27485-25G 27485-500MG 27485-5G
76939-126EA 206.88 USD
76939-126 76939-128 76939-130 76939-132
6-Bromo-2-chloronicotinaldehyde ≥97%
6-Bromo-2-chloronicotinaldehyde
Formula: C₆H₃BrClNO
Storage Temperature: Refrigerator
MDL Number: MFCD11857709
CAS Number: 1125410-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR