5-Methoxymethyl-thiophene ≥97%

Supplier: AOBChem USA

Warning

27338-25G 27338-5G
76939-094EA 346.92 USD
76939-094 76939-096
5-Methoxymethyl-thiophene ≥97%
5-Methoxymethyl-thiophene

Precaución: for Research Use Only

Formula: C₆H₈OS
MW: 128.192 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22054632
CAS Number: 1487-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR