6-Pyrrolidin-1-ylnicotinaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 6-(1-Pyrrolidinyl)nicotinaldehyde

27293-1G 27293-25G 27293-5G
76939-082EA 99.73 USD
76939-082 76939-084 76939-086
6-Pyrrolidin-1-ylnicotinaldehyde ≥97% research grade
6-Pyrrolidin-1-ylnicotinaldehyde
Formula: C₁₀H₁₂N₂O
MW: 176.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06200881
CAS Number: 261715-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR