2-Fluoro-3-(methoxycarbonyl)benzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-3-(methoxycarbonyl)phenylboronic acid pinacol ester, Methyl 2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

27286-1G 27286-25G 27286-500MG 27286-5G
76939-066EA 203.7 USD
76939-066 76939-068 76939-070 76939-072
2-Fluoro-3-(methoxycarbonyl)benzeneboronic acid pinacol ester ≥97%
2-Fluoro-3-(methoxycarbonyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BFO₄
MW: 280.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16996311
CAS Number: 1638847-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR