Ethyl 3-bromo-5-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

27258-1G 27258-25G 27258-5G
76939-058EA 79.57 USD
76939-058 76939-060 76939-062
Ethyl 3-bromo-5-fluorobenzoate ≥97%
Ethyl 3-bromo-5-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MW: 247.061 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10566820
CAS Number: 353743-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR