Methyl 5-methoxy-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

27177-25G 27177-5G
76939-048EA 346.92 USD
76939-048 76939-050
Methyl 5-methoxy-2-methylbenzoate ≥97%
Methyl 5-methoxy-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD11045607
CAS Number: 73502-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR