5-Methoxy-2-Methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

26911-1G 26911-25G 26911-500MG 26911-5G
76938-994EA 352.22 USD
76938-994 76938-996 76938-998 76939-000
5-Methoxy-2-Methylphenylboronic acid pinacol ester ≥97%
5-Methoxy-2-Methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₁BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD28156175
CAS Number: 1641570-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR