N-(2-Methoxy-5-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-Yl)Pyridin-3-Yl)Methanesulfonamide ≥95%
Supplier: AOBChem USA
|
Warning
|
26782-1G
26782-25G
26782-500MG
26782-5G
76938-978EA
379.81
USD
76938-978
76938-980
76938-982
76938-984
N-(2-Methoxy-5-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-Yl)Pyridin-3-Yl)Methanesulfonamide ≥95%
N-(2-Methoxy-5-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-Yl)Pyridin-3-Yl)Methanesulfonamide
Precaución: for Research Use Only
Formula:
C₁₃H₂₁BN₂O₅S MW: 328.192 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12964555 CAS Number: 1083326-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...