Ethyl-5-bromo-2-chloroisonicotinate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 5-bromo-2-chloroisonicotinate

26551-25G 26551-5G
76938-932EA 275.84 USD
76938-932 76938-934
Ethyl-5-bromo-2-chloroisonicotinate ≥95%
Ethyl-5-bromo-2-chloroisonicotinate
Formula: C₈H₇BrClNO₂
MW: 264.51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698168
CAS Number: 1214346-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR