6-Bromo-2-fluoro-3-methoxybenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
26525-1G
26525-25G
26525-5G
76938-922EA
117.76
USD
76938-922
76938-924
76938-926
6-Bromo-2-fluoro-3-methoxybenzoic acid ≥97%
6-Bromo-2-fluoro-3-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₆BrFO₃ MW: 249.034 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09261247 CAS Number: 935534-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...