6-Bromo-2-chloro-3-methylphenol ≥97% research grade

Supplier: AOBChem USA

Warning

26511-1G 26511-25G 26511-5G
76938-912EA 99.73 USD
76938-912 76938-914 76938-916
6-Bromo-2-chloro-3-methylphenol ≥97% research grade
6-Bromo-2-chloro-3-methylphenol
Formula: C₇H₆BrClO
Storage Temperature: Refrigerator
MDL Number: MFCD16295099
CAS Number: 1226808-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR