5-Methoxy-1H-pyrazolo[3,4-b]pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
26439-1G
26439-500MG
26439-5G
76938-878EA
558.04
USD
76938-878
76938-880
76938-882
5-Methoxy-1H-pyrazolo[3,4-b]pyridine ≥97%
5-Methoxy-1H-pyrazolo[3,4-b]pyridine
Precaución: for Research Use Only
Formula:
C₇H₇N₃O MW: 149.15 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18254575 CAS Number: 1256804-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...