6-Bromobenzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

26416-25G 26416-5G
76938-866EA 318.27 USD
76938-866 76938-868
6-Bromobenzo[b]thiophene ≥95%
6-Bromobenzo[b]thiophene

Precaución: for Research Use Only

Formula: C₈H₅BrS
Storage Temperature: Ambient
MDL Number: MFCD11226847
CAS Number: 17347-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR