O-3-iodophenyl N,N-diethylcarbamate ≥97%

Supplier: AOBChem USA

Warning

26353-1G 26353-25G 26353-5G
76938-856EA 88.06 USD
76938-856 76938-858 76938-860
O-3-iodophenyl N,N-diethylcarbamate ≥97%
O-3-iodophenyl N,N-diethylcarbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₄INO₂
Storage Temperature: Ambient
CAS Number: 863870-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR