Methyl-3-bromo-5-hydroxybenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 3-bromo-5-hydroxybenzoate

26323-25G 26323-5G
76938-844EA 265.23 USD
76938-844 76938-846
Methyl-3-bromo-5-hydroxybenzoate ≥97%
Methyl-3-bromo-5-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
MW: 231.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10566800
CAS Number: 192810-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR