5-Iodo-2-methoxy-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

26243-1G 26243-25G 26243-5G
76938-830EA 82.76 USD
76938-830 76938-832 76938-834
5-Iodo-2-methoxy-4-methylpyridine ≥97%
5-Iodo-2-methoxy-4-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₈INO
MW: 249.049 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26045268
CAS Number: 1092978-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR