6-Chloro-1H-indazole-4-boronic acid ≥95%

Supplier: AOBChem USA

Warning

26116-1G 26116-25G 26116-500MG 26116-5G
76938-786EA 350.1 USD
76938-786 76938-788 76938-790 76938-792
6-Chloro-1H-indazole-4-boronic acid ≥95%
6-Chloro-1H-indazole-4-boronic acid

Precaución: for Research Use Only

Formula: C₇H₆BClN₂O₂
MW: 196.399 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26045254
CAS Number: 2121512-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR