8-Fluoroisoquinoline ≥97%

Supplier: AOBChem USA

Warning

26093-1G 26093-25G 26093-5G
76938-772EA 114.58 USD
76938-772 76938-774 76938-776
8-Fluoroisoquinoline ≥97%
8-Fluoroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₆FN
MW: 147.1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878369
CAS Number: 1075-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR