Methyl 6-bromo-2-fluoro-3-methylbenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
26022-25G
26022-5G
76938-760EA
595.17
USD
76938-760
76938-762
Methyl 6-bromo-2-fluoro-3-methylbenzoate ≥97%
Methyl 6-bromo-2-fluoro-3-methylbenzoate
Formula:
C₉H₈BrFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD24848718 CAS Number: 1437780-03-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...