Methyl 2-chloro-3-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

25938-25G 25938-5G
76938-712EA 595.17 USD
76938-712 76938-714
Methyl 2-chloro-3-(trifluoromethyl)benzoate ≥97%
Methyl 2-chloro-3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD06203770
CAS Number: 378231-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR