5-Methoxy-2-phenylbenzonitrile ≥97%

Supplier: AOBChem USA

Warning

25867-1G 25867-25G 25867-500MG 25867-5G
76938-698EA 215.37 USD
76938-698 76938-700 76938-702 76938-704
5-Methoxy-2-phenylbenzonitrile ≥97%
5-Methoxy-2-phenylbenzonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₂
MW: 212.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039098
CAS Number: 934691-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR