Isoquinoline-5-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

25740-1G 25740-25G 25740-500MG 25740-5G
76938-662EA 171.87 USD
76938-662 76938-664 76938-666 76938-668
Isoquinoline-5-boronic acid pinacol ester ≥97%
Isoquinoline-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₁₈BNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD23331387
CAS Number: 1338914-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR