tert-Butyl (2-bromophenyl)carbamate ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: N-(tert-Butoxycarbonyl)-2-bromoaniline

25641-25G 25641-5G
76938-628EA 204.76 USD
76938-628 76938-630
tert-Butyl (2-bromophenyl)carbamate ≥97% research grade
tert-Butyl (2-bromophenyl)carbamate
Formula: C₁₁H₁₄BrNO₂
MW: 272.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01006614
CAS Number: 78839-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR