Methyl 4-chloro-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

25449-25G 25449-5G
76938-616EA 198.39 USD
76938-616 76938-618
Methyl 4-chloro-3-methoxybenzoate ≥97%
Methyl 4-chloro-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MW: 200.619 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683861
CAS Number: 116022-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR