7-Bromo-4-fluoro-1H-indazole ≥97% research grade

Supplier: AOBChem USA

Warning

25408-1G 25408-25G 25408-500MG 25408-5G
76938-600EA 198.39 USD
76938-600 76938-602 76938-604 76938-606
7-Bromo-4-fluoro-1H-indazole ≥97% research grade
7-Bromo-4-fluoro-1H-indazole
Formula: C₇H₄BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD09749914
CAS Number: 1000341-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR