6-Bromo-2-methylpyridine-3-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

25118-1G 25118-25G 25118-500MG 25118-5G
76938-566EA 260.99 USD
76938-566 76938-568 76938-570 76938-572
6-Bromo-2-methylpyridine-3-carboxylic acid ≥95%
6-Bromo-2-methylpyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.032 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13188658
CAS Number: 1060805-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR