N-Boc-4-bromo-3-fluoroaniline ≥95%

Supplier: AOBChem USA

Warning

24967-1G 24967-25G 24967-500MG 24967-5G
76938-540EA 138.98 USD
76938-540 76938-542 76938-544 76938-546
N-Boc-4-bromo-3-fluoroaniline ≥95%
N-Boc-4-bromo-3-fluoroaniline
Formula: C₁₁H₁₃BrFNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11976284
CAS Number: 868735-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR