Methyl-2-chloro-6-methoxynicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 2-chloro-6-methoxynicatinate, Methyl 2-chloro-6-methoxynicotinate

24864-1G 24864-25G 24864-5G
76938-528EA 62.6 USD
76938-528 76938-530 76938-532
Methyl-2-chloro-6-methoxynicotinate ≥97%
Methyl-2-chloro-6-methoxynicotinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD08275100
CAS Number: 95652-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR