7-Chloro-1H-indazole-4-boronic acid ≥95%

Supplier: AOBChem USA

Warning

24817-1G 24817-500MG 24817-5G
76938-514EA 468.92 USD
76938-514 76938-516 76938-518
7-Chloro-1H-indazole-4-boronic acid ≥95%
7-Chloro-1H-indazole-4-boronic acid

Precaución: for Research Use Only

Formula: C₇H₆BClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD22628222
CAS Number: 1451393-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR