6-Bromo-1-chloro-2-(methoxymethoxy)naphthalene ≥97%

Supplier: AOBChem USA

Warning

24649-1G 24649-500MG 24649-5G
76938-502EA 448.77 USD
76938-502 76938-504 76938-506
6-Bromo-1-chloro-2-(methoxymethoxy)naphthalene ≥97%
6-Bromo-1-chloro-2-(methoxymethoxy)naphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD28977151
CAS Number: 2179038-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR