Methyl-2,3-dichloroisonicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 2,3-dichloroisonicotinate

24435-1G 24435-25G 24435-5G
76938-488EA 62.6 USD
76938-488 76938-490 76938-492
Methyl-2,3-dichloroisonicotinate ≥97%
Methyl 2,3-dichloroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MW: 206.0300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09951946
CAS Number: 603124-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR