Methyl 2-bromo-5-chloroisonicotinate ≥95%

Supplier: AOBChem USA

Warning

24264-1G 24264-25G 24264-5G
76938-458EA 62.6 USD
76938-458 76938-460 76938-462
Methyl 2-bromo-5-chloroisonicotinate ≥95%
Methyl 2-bromo-5-chloroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₅BrClNO₂
MW: 250.477 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698101
CAS Number: 1214336-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR