5-Iodo-2-methylphenol ≥97%

Supplier: AOBChem USA

Warning

24201-1G 24201-25G 24201-500MG 24201-5G
76938-442EA 126.25 USD
76938-442 76938-444 76938-446 76938-448
5-Iodo-2-methylphenol ≥97%
5-Iodo-2-methylphenol
Formula: C₇H₇IO
MW: 234.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12923189
CAS Number: 183803-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR