6-Isopropoxypyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

24169-1G 24169-25G 24169-500MG 24169-5G
76938-426EA 260.99 USD
76938-426 76938-428 76938-430 76938-432
6-Isopropoxypyridin-3-ol ≥97%
6-Isopropoxypyridin-3-ol

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16997398
CAS Number: 903886-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR