Isopropyl 5-bromonicotinamide ≥97%

Supplier: AOBChem USA

Warning

24075-1G 24075-25G 24075-5G
76938-388EA 64.72 USD
76938-388 76938-390 76938-392
Isopropyl 5-bromonicotinamide ≥97%
Isopropyl 5-bromonicotinamide

Precaución: for Research Use Only

Formula: C₉H₁₁BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD06016757
CAS Number: 104290-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR