6-Phenoxynicotinic acid ≥97%

Supplier: AOBChem USA

Warning

23982-25G 23982-5G
76938-358EA 381.93 USD
76938-358 76938-360
6-Phenoxynicotinic acid ≥97%
6-Phenoxynicotinic acid

Precaución: for Research Use Only

Formula: C₁₂H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD02682068
CAS Number: 51362-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR