5-Methyl-2-phenylpyridine ≥97%

Supplier: AOBChem USA

Synonyms: 6-Phenyl-3-picoline

23713-1G 23713-25G 23713-5G
76938-312EA 106.09 USD
76938-312 76938-314 76938-316
5-Methyl-2-phenylpyridine ≥97%
5-Methyl-2-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₁N
MW: 169.23
MDL Number: MFCD04114180
CAS Number: 27012-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR