N,N-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 6-(Dimethylamino)pyridine-3-boronic acid pinacol ester

23661-1G 23661-25G 23661-5G
76938-302EA 70.02 USD
76938-302 76938-304 76938-306
N,N-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine ≥97% research grade
N,N-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Formula: C₁₃H₂₁BN₂O₂
MW: 248.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11975406
CAS Number: 1036991-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR