6-Ethoxy-2-fluoro-3-methylphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

23336-25G 23336-5G
76938-270EA 573.95 USD
76938-270 76938-272
6-Ethoxy-2-fluoro-3-methylphenylboronic acid ≥97% research grade
6-Ethoxy-2-fluoro-3-methylphenylboronic acid
Formula: C₉H₁₂BFO₃
Storage Temperature: Ambient
MDL Number: MFCD19981512
CAS Number: 1451391-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR