5-Methyl-1H-indazole-4-boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (5-Methyl-1H-indazol-4-yl)boronic acid

23266-1G 23266-25G 23266-5G
76938-248EA 62.6 USD
76938-248 76938-250 76938-252
5-Methyl-1H-indazole-4-boronic acid ≥97%
5-Methyl-1H-indazole-4-boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₂
MW: 175.9800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10001490
CAS Number: 1245816-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR